missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2’,3’,5’-Tri-O-acetyl Guanosine-13D2,15N, TRC
Specifications
Specifications
| Chemical Name or Material | 2’,3’,5’-Tri-O-acetyl Guanosine-13C2,15N |
| Molecular Formula | 13C2 C14 H19 15N N4 O8 |
| Quantity | 1 mg |
| Synonym | 2’,3’,5’-Tri-O-acetylguanosine-13C2,15N, Guanosine Triacetate-13C2,15N, NSC 66387-13C2,15N, Triacetylguanosine-13C2,15N |
| InChI Formula | InChI=1S/C16H19N5O8/c1-6(22)26-4-9-11(27-7(2)23)12(28-8(3)24)15(29-9)21-5-18-10-13(21)19-16(17)20-14(10)25/h5,9,11-12,15H,4H2,1-3H3,(H3,17,19,20,25)/t9-,11-,12-,15-/m1/s1/i10+1,13+1,18+1 |
| SMILES | CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)n2c[15n][13c]3[13c]2N=C(N)NC3=O |
| IUPAC Name | [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(2-amino-6-oxo-1H-purin-9-yl)oxolan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 412.329 |
| Formula Weight | 412.127 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?