Learn More
2,3-Dihydro-5H-1,4-benzodioxepin-5-one, 98%
Product Code. 11343157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343157 5 g 5g
11333157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11343157 Supplier Thermo Scientific Alfa Aesar Supplier No. L13582.06
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
CAS: 22891-52-7 | C9H8O3 | 164.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22891-52-7
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00236035
InChI Key PYVNMBGWXYJCCL-UHFFFAOYSA-N
Synonym 2,3-dihydro-5h-1,4-benzodioxepin-5-one, 2h-benzo e 1,4 dioxepin-5 3h-one, 2,3-dihydro-5h-1,4-benzodioxipin-5-one, 5h-1,4-benzodioxepin-5-one, 2,3-dihydro, 4-19-00-01699 beilstein handbook reference, 2h,3h-benzo f 1,4-dioxepin-5-one
PubChem CID 211314
IUPAC Name 2,3-dihydro-1,4-benzodioxepin-5-one
SMILES C1COC(=O)C2=CC=CC=C2O1
Specifications
Melting Point 29°C to 31°C
Boiling Point 178°C to 184°C (15 mmHg)
Refractive Index 1.558
Quantity 5 g
Beilstein 4464
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 2,3-Dihydro-5H-1,4-benzodioxepin-5-one