missing translation for 'onlineSavingsMsg'
Learn More

2,4'-(2-Pyridinyl-2methylene)diphenol, TRC

Product Code. 30180274
Change view
Click to view available options
Quantity:
25 mg
500 mg
Unit Size:
25mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180274 500 mg 500mg
30180056 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180274 Supplier Toronto Research Chemicals Supplier No. TRCP991840500MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16985-05-0
Molecular Formula C18 H15 N O2
Molecular Weight (g/mol) 277.32
Synonym 2,4'-[(Pyridin-2-yl)methylene]bisphenol, 2-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenol, Bisacodyl Imp. B (EP)
IUPAC Name 2-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenol
SMILES Oc1ccc(cc1)C(c2ccccn2)c3ccccc3O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H15NO2/c20-14-10-8-13(9-11-14)18(16-6-3-4-12-19-16)15-5-1-2-7-17(15)21/h1-12,18,20-21H
Formula Weight 277.1103
Chemical Name or Material 2-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.