missing translation for 'onlineSavingsMsg'
Learn More

2,4,7,9-Tetramethyl-5-decyne-4,7-diol-d10, TRC

Product Code. 30425647
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425647 25 mg 25mg
30410686 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30425647 Supplier Toronto Research Chemicals Supplier No. TRCT30250225MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,7,9-Tetramethyl-5-decyne-4,7-diol-d10
Molecular Formula C14 D10 H16 O2
Quantity 25 mg
Synonym 1,4-Diisobutyl-1,4-dimethylbutynediol-d10, 2,4,7,9-Tetramethyl-5-decyn-4,7-diol-d10, Acetylenol E 00-d10, NSC 5630-d10, Olfine AK 02-d10, Olfine PD 301-d10, Olfine STG-E-d10, PD 301-d10, Surfynol 104-d10, Surfynol 104A-d10, Surfynol 104BC-d10, Surfynol 104E-d10, Surfynol 104H-d10, Surfynol 104PA-d10, Surfynol CT 111-d10, Surfynol PG 50-d10, Surfynol TG-d10, Zetasperse 1200-d10
InChI Formula InChI=1S/C14H26O2/c1-11(2)9-13(5,15)7-8-14(6,16)10-12(3)4/h11-12,15-16H,9-10H2,1-6H3/i5D3,6D3,9D2,10D2
SMILES [2H]C([2H])([2H])C(O)(C#CC(O)(C([2H])([2H])[2H])C([2H])([2H])C(C)C)C([2H])([2H])C(C)C
IUPAC Name 3,3,8,8-tetradeuterio-2,9-dimethyl-4,7-bis(trideuteriomethyl)dec-5-yne-4,7-diol
Molecular Weight (g/mol) 236.417
Formula Weight 236.256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.