missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoyl chloride, 97%

Product Code. 11489036
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489036 5 g 5g
11499036 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489036 Supplier Thermo Scientific Alfa Aesar Supplier No. A17920.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 72482-64-5 | C7H3ClF2O | 176.55 g/mol

2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72482-64-5
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD00000658
InChI Key JSWRVDNTKPAJLB-UHFFFAOYSA-N
PubChem CID 588081
IUPAC Name 2,4-difluorobenzoyl chloride
SMILES FC1=CC=C(C(Cl)=O)C(F)=C1

Specifications

Density 1.437
Boiling Point 96°C to 98°C (15 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.5145
Quantity 5 g
UN Number UN3265
Beilstein 4800566
Sensitivity Moisture sensitive
Synonym 2,4-difluorobenzoylchloride, benzoyl chloride, 2,4-difluoro, benzoyl chloride,2,4-difluoro, acmc-1bfet, 2,4difluorobenzoylchloride, 2,4-difluorobezoyl chloride, 2,4 difluorobenzoyl chloride, 2,4-difluoro benzoylchloride, 2,4-difluoro-benzoylchloride, 2,4-difluoro-benzoyl chloride
Solubility Information Reacts with water.
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,4-Difluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.