missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diethoxyaniline, 96%, Thermo Scientific™

Product Code. 11306886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306886 1 g 1g
11316886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306886 Supplier Thermo Scientific Alfa Aesar Supplier No. L04296.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-85-9
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD00015144
InChI Key XPKFTIYOZUJAGA-UHFFFAOYSA-N
Synonym benzenamine, 2,5-diethoxy, aminohydroquinone diethyl ether, 2,5-diethoxy aniline, aniline, 2,5-diethoxy, 2,5-diethoxyphenyl amine, 2,5-diethoxyphenylamine, 2.5-diethoxyaniline, aniline,5-diethoxy, 2,5-diathoxy-anilin, 2,5-diethoxybenzenamine
PubChem CID 66756
IUPAC Name 2,5-diethoxyaniline
SMILES CCOC1=CC(=C(C=C1)OCC)N

Specifications

Melting Point 84°C to 86°C
Boiling Point 190°C to 192°C (30 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 880466
Formula Weight 181.24
Percent Purity 96%
Chemical Name or Material 2,5-Diethoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.