missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorodihydrofluorescein Diacetate, TRC

Product Code. 17914765
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914765 50 mg 50mg
17960565 100 mg 100mg
17921595 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17914765 Supplier Toronto Research Chemicals Supplier No. TRCD43421550MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4091-99-0
Molecular Formula C24H16Cl2O7
Molecular Weight (g/mol) 487.29
Synonym 2-[3,6-Bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic Acid, o-(2,7-Dichloro-3,6-dihydroxyxanthen-9-yl)benzoic Acid Diacetate, 2',7'-Dichlorodihydrofluorescein Diacetate, DCFH, H2DCFDA;
SMILES CC(=O)OC1=C(C=C2C(=C1)OC3=CC(=C(C=C3C2C4=CC=CC=C4C(=O)O)Cl)OC(=O)C)Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H16Cl2O7/c1-11(27)31-21-9-19-15(7-17(21)25)23(13-5-3-4-6-14(13)24(29)30)16-8-18(26)22(32-12(2)28)10-20(16)33-19/h3-10,23H,1-2H3,(H,29,30)
Formula Weight 486.0273
Chemical Name or Material 2',7'-Dichlorodihydrofluorescein Diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.