missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzeneboronic acid, 97%

Product Code. 11342419
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342419 25 g 25g
11332419 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342419 Supplier Thermo Scientific Alfa Aesar Supplier No. H27327.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 308103-40-4 | C8H9BO3 | 163.97 g/mol

2-Acetylphenylboronic acid is a reactant used for a variety of coupling reactions, as well as synthesis and hydroxy masking, Suzuki-Miyaura cross-coupling reactions, rhodium-catalyzed annulation of ynamides, transient masking of hydroxy groups, cationic palladium complex-catalyzed diastereoselective tandem annulation, hydrogen peroxide mediated formation of heteroaryl ethers and copper-catalyzed transmetalation and homocoupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylphenylboronic acid is a reactant used for a variety of coupling reactions, as well as synthesis and hydroxy masking, Suzuki-Miyaura cross-coupling reactions, rhodium-catalyzed annulation of ynamides, transient masking of hydroxy groups, cationic palladium complex-catalyzed diastereoselective tandem annulation, hydrogen peroxide mediated formation of heteroaryl ethers and copper-catalyzed transmetalation and homocoupling.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308103-40-4
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01321263
InChI Key ZKAOVABYLXQUTI-UHFFFAOYSA-N
Synonym 2-acetylphenyl boronic acid, boronic acid, 2-acetylphenyl, 2'-boronoacetophenone, 1-2-boronophenyl ethan-1-one, 2-acetylbenzeneboronic acid, 2-acetylbenzenboronicacid, 1-2-dihydroxyboranyl phenyl ethan-1-one, 2-acetylphenylboronicacid, pubchem5373, acmc-1agh6
PubChem CID 2734309
IUPAC Name (2-acetylphenyl)boronic acid
SMILES CC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point ∽170°C (decomposition)
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 163.97
Percent Purity 97%
Chemical Name or Material 2-Acetylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.