Learn More
2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 95%
Beschreibung
2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Spezifikation
Spezifikation
| Quantity | 1 g |
| Beilstein | 15519102 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 288.07 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester |