missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzothiazole (Tech.), 90%, ACROS Organics™

Product Code. 10008651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10008651 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10008651 Supplier Thermo Scientific Acros Supplier No. 103940050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1747-60-0
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C8H8N2OS
Quantity 5 g
Synonym 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393
InChI Key KZHGPDSVHSDCMX-UHFFFAOYSA-N
SMILES COC1=CC2=C(C=C1)N=C(S2)N
IUPAC Name 6-methoxy-1,3-benzothiazol-2-amine
Molecular Weight (g/mol) 180.225
PubChem CID 15630
Formula Weight 180.23
Grade Technical
Chemical Name or Material 2-Amino-6-methoxybenzothiazole, tech., 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.