missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)thiazole, 97%

Product Code. 15444808
Change view
Click to view available options
Quantity:
5 g
30 g
Unit Size:
30g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444808 5 g 5g
15454808 30 g 30g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444808 Supplier Thermo Scientific Alfa Aesar Supplier No. H58561.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 170961-15-6 | C8H12N2O2S | 200.256 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170961-15-6
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.256
MDL Number MFCD09038327
InChI Key NCJXQSNROJRSSL-UHFFFAOYSA-N
Synonym tert-butyl thiazol-2-ylcarbamate, tert-butyl n-1,3-thiazol-2-yl carbamate, n-boc-2-aminothiazole, 2-boc-amino thiazole, 2-tert-butoxycarbonylamino thiazole, thiazol-2-yl-carbamic acid tert-butyl ester, pubchem23669, tertbutylthiazolylcarbamate, 2-n-boc-amino thiazole
PubChem CID 11858898
IUPAC Name tert-butyl N-(1,3-thiazol-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=CS1

Specifications

Melting Point 180°C to 185°C
Quantity 5 g
Formula Weight 200.25
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)thiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.