Learn More
2'-Bromo-4'-fluoroacetophenone, 98%
Product Code. 11344236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344236 1 g 1g
11354236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11344236 Supplier Thermo Scientific Alfa Aesar Supplier No. L19320.03
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
CAS: 1006-39-9 | C8H6BrFO | 217.04 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1006-39-9
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD00077464
InChI Key RCXFSBRMWBFWMH-UHFFFAOYSA-N
Synonym 2'-bromo-4'-fluoroacetophenone, 1-2-bromo-4-fluorophenyl ethanone, 1-acetyl-2-bromo-4-fluorobenzene, 1-2-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-4-fluorophenyl, 1-2-bromo-4-fluoro-phenyl ethanone, pubchem8533, acmc-2097rj, ksc499c4r, timtec-bb sbb006561
PubChem CID 2736304
IUPAC Name 1-(2-bromo-4-fluorophenyl)ethanone
SMILES CC(=O)C1=C(Br)C=C(F)C=C1
Specifications
Boiling Point 150°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.549
Quantity 1 g
Beilstein 2249797
Formula Weight 217.04
Percent Purity 98%
Chemical Name or Material 2'-Bromo-4'-fluoroacetophenone