missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium Chloride (90%), TRC

Product Code. 30358094
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358094 1 g -
30368231 10 g -
30353067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358094 Supplier Toronto Research Chemicals Supplier No. TRCC3656951G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37091-73-9
Molecular Formula C5H10Cl2N2
Molecular Weight (g/mol) 169.05
Synonym 2-Chloro-4,5-dihydro-1,3-dimethyl-1H-imidazolium Chloride, 2-Chloro-1,3-dimethyl-4,5-dihydroimidazolium Chloride
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
SMILES [Cl-].CN1CC[N+](=C1Cl)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H10ClN2.ClH/c1-7-3-4-8(2)5(7)6;/h3-4H2,1-2H3;1H/q+1;/p-1
Formula Weight 168.0221
Chemical Name or Material 2-Chloro-1,3-dimethylimidazolinium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.