missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxybenzaldehyde, 97%

Product Code. 10041264 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10041264 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10041264 Supplier Thermo Scientific Acros Supplier No. 398030010
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56962-11-9 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56962-11-9
Melting Point 145°C to 148°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H5ClO2
Linear Formula ClC6H3(OH)CHO
Quantity 1 g
Synonym benzaldehyde, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzaldehyde, benzaldehyde, chloro-4-hydroxy, 2-chloro-4-hydroxybenzaldehyde, pubchem7366, acmc-209lv6, ksc497o6r, 2-chlor-4-hydroxy-benzaldehyd, 2-chloranyl-4-oxidanyl-benzaldehyde
InChI Key ZMOMCILMBYEGLD-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1O)Cl)C=O
IUPAC Name 2-chloro-4-hydroxybenzaldehyde
Molecular Weight (g/mol) 156.57
PubChem CID 185363
Formula Weight 156.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-hydroxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-hydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.