missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, TRC

Product Code. 30354551
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354551 1 g -
30349723 10 g 10g
30364251 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354551 Supplier Toronto Research Chemicals Supplier No. TRCC5856081G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013916-37-4
Molecular Formula C13H14ClN3O
Molecular Weight (g/mol) 263.72
Synonym 2-Chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one, 2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one
IUPAC Name 2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
SMILES CC1=CC(=O)N(C2CCCC2)c3nc(Cl)ncc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H14ClN3O/c1-8-6-11(18)17(9-4-2-3-5-9)12-10(8)7-15-13(14)16-12/h6-7,9H,2-5H2,1H3
Chemical Name or Material 2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.