missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroaniline-d6, CDN
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | 2-Chloroaniline-D6 |
| CAS | 1276197-37-5 |
| Molecular Formula | C6 D6 Cl N |
| Quantity | 0.05 g |
| Synonym | 2-Chloroaniline-d6, 2-Chloroaniline-N,N,3,4,5,6-d6, 2-Chlorobenzenamine-N,N,3,4,5,6-d6, 6-Chlorobenzenamine-N,N,2,3,4,5-d6 |
| InChI Formula | InChI=1 S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4 H,8H2/i1D,2 D,3 D,4 D/hD2 |
| SMILES | [2 H]N([2 H])c1c([2 H])c([2 H])c([2 H])c([2 H])c1Cl |
| IUPAC Name | 2-chloro-N,N,3,4,5,6-hexadeuterioaniline |
| Molecular Weight (g/mol) | 133.6085 |
| Formula Weight | 133.0565 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction