missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoylacetonitrile, 98%, ACROS Organics™

Product Code. 10763783 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10763783 1 g Glass bottle 1g
10773783 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763783 Supplier Thermo Scientific Acros Supplier No. 355430010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40018-25-5
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051624
InChI Key SBSWHTFHLWSSQS-UHFFFAOYSA-N
Synonym 2-chlorobenzoylacetonitrile, 3-2-chlorophenyl-3-oxopropanenitrile, 2-chlorobenzoyl acetonitrile, pubchem12047, acmc-1arra, 0-chlorocyanoacetophenone, o-chlorobenzoyl acetonitrile, 2'-chloro-2-cyanoacetophenone, timtec-bb sbb019129, 2-chlorobenzoylacetonitirle
PubChem CID 2734201
IUPAC Name 3-(2-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC=C(C(=C1)C(=O)CC#N)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55°C to 59°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 179.61
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzoylacetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.