missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl alcohol, 98%

Product Code. 11472517
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472517 10 g 10g
11482517 50 g 50g
11339619 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472517 Supplier Thermo Scientific Alfa Aesar Supplier No. A14791.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 446-51-5 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-51-5
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004601
InChI Key QEHXDOJPVIHUDO-UHFFFAOYSA-N
Synonym 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8
PubChem CID 67969
IUPAC Name (2-fluorophenyl)methanol
SMILES OCC1=CC=CC=C1F

Specifications

Density 1.175
Boiling Point 201°C
Flash Point 90°C (194°F)
Odor Characteristic
Refractive Index 1.514
Quantity 10 g
Beilstein 2079486
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.