missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroisobutyric acid, 95%

Product Code. 11495560
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495560 1 g 1g
15413317 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495560 Supplier Thermo Scientific Alfa Aesar Supplier No. H25837.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 63812-15-7 | C4H7FO2 | 106.096 g/mol

2-Fluoroisobutyric acid chloride was synthesized from 2-fluoroisobutyric acid and oxalyl chloride. Reactant in the synthesis of 2-Fluoro-Acids, Esters, and amides From a-Dicyanoepoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoroisobutyric acid chloride was synthesized from 2-fluoroisobutyric acid and oxalyl chloride. Reactant in the synthesis of 2-Fluoro-Acids, Esters, and amides From a-Dicyanoepoxides.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63812-15-7
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.096
MDL Number MFCD03452788
InChI Key NZDOWZQRNZLBOY-UHFFFAOYSA-N
Synonym 2-fluoroisobutyric acid, 2-fluoroisobutanoic acid, 2-fluoro-2-methyl-propanoic acid, 2-fluoro-2-methylpropionic acid, fluoroisobutyric acid, acmc-1b6kg, 2-methyl-2-fluoropropionic acid, 2-fluoro-2-methyl-propionic acid, 2-fluoranyl-2-methyl-propanoic acid, propanoic acid, 2-fluoro-2-methyl
PubChem CID 2782828
IUPAC Name 2-fluoro-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)F

Specifications

Boiling Point 57°C to 58°C (10 mmHg)
Refractive Index 1.385
Quantity 1 g
UN Number UN2922
Solubility Information Soluble in water.
Formula Weight 106.1
Percent Purity 95%
Chemical Name or Material 2-Fluoroisobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.