Learn More
2-Hydroxy-4-methylpyrimidine hydrochloride, 97%, Thermo Scientific™
Product Code. 11455906
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455906 100 g 100g
11445906 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11455906 Supplier Thermo Scientific Alfa Aesar Supplier No. B22255.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5348-51-6
Molecular Formula C5H7ClN2O
Molecular Weight (g/mol) 146.574
MDL Number MFCD00151347
InChI Key FDRYQDWXFZBBAD-UHFFFAOYSA-N
Synonym 4-methylpyrimidin-2-ol hydrochloride, 2-hydroxy-4-methylpyrimidine hydrochloride, 2-hydroxy-4-methylpyrimidine hcl, 6-methylpyrimidin-2 1h-one hydrochloride, 2-hydroxy-4-methylpyrimidine hydro-chloride, 2 1h-pyrimidinone, 4-methyl-, monohydrochloride, 4-methyl-3-hydropyrimidin-2-one, chloride, 2-hydroxy-4-methylpyrimidinehydrochloride, zlchem 746, pubchem22005
PubChem CID 2723648
IUPAC Name 6-methyl-1H-pyrimidin-2-one;hydrochloride
SMILES CC1=CC=NC(=O)N1.Cl
Specifications
Melting Point ∼240°C (decomposition)
Odor Odorless
Quantity 100 g
Beilstein 3691454
Formula Weight 146.58
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4-methylpyrimidine hydrochloride