missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol Cell Culture MP Biomedicals

Product Code. 11471921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471921 250 mL 250mL
11481921 500 mL 500mL
11491921 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471921 Supplier MP Biomedicals Supplier No. 0219470583
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
IUPAC Name 2-sulfanylethan-1-ol
SMILES OCCS

Specifications

Density 1.12g/mL
Assay Percent Range >98%
Quantity 250 mL
Formula Weight 78.13
Grade Cell Culture Reagent
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol, Molecular Biology and Proteomics
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Mercaptoethanol, cell culture reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.