missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine, 98%

Product Code. 11460284
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460284 500 g 500g
11440284 25 g 25g
11450284 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460284 Supplier Thermo Scientific Alfa Aesar Supplier No. A13675.36
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1628-89-3 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1628-89-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006262
InChI Key IWTFOFMTUOBLHG-UHFFFAOYSA-N
Synonym pyridine, 2-methoxy, 2-methoxy pyridine, pyridine, methoxy, 2-methoxy-pyridine, unii-geb38j8108, methyl 2-pyridyl ether, methoxypyridine, 6-methoxypyridine, methyl pyridinyl ether, pubchem13201
PubChem CID 74201
IUPAC Name 2-methoxypyridine
SMILES COC1=CC=CC=N1

Specifications

Density 1.046
Boiling Point 142°C to 143°C
Flash Point 32°C (89°F)
Refractive Index 1.504
Quantity 500 g
UN Number UN3271
Beilstein 108189
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Methoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.