missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[Methyl-1-(4-methoxyphenyl)methoxy]propyl-4’-nitrophenyl-d6 Carbonate, TRC
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Specifications
Specifications
| Chemical Name or Material | 2-[Methyl-1-(4-methoxyphenyl)methoxy]propyl-4’-nitrophenyl-d6 Carbonate |
| Molecular Formula | C19 D6 H15 N O7 |
| Quantity | 10 mg |
| Synonym | 2-[(4-Methoxyphenyl)methoxy]-1,1-dimethylethyl-carbonic Acid-d6 4-Nitrophenyl Ester |
| InChI Formula | InChI=1S/C19H21NO7/c1-19(2,13-25-12-14-4-8-16(24-3)9-5-14)27-18(21)26-17-10-6-15(7-11-17)20(22)23/h4-11H,12-13H2,1-3H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])C(COCc1ccc(OC)cc1)(OC(=O)Oc2ccc(cc2)[N+](=O)[O-])C([2H])([2H])[2H] |
| IUPAC Name | [1,1,1,3,3,3-hexadeuterio-2-[(4-methoxyphenyl)methoxymethyl]propan-2-yl] (4-nitrophenyl) carbonate |
| Molecular Weight (g/mol) | 381.409 |
| Formula Weight | 381.169 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction