missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-phenylsuccinimide, 99%

Product Code. p-7033860
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427927 1 g 1g
15437927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427927 Supplier Thermo Scientific Alfa Aesar Supplier No. H51038.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1497-17-2 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1497-17-2
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005496
InChI Key UDESUGJZUFALAM-UHFFFAOYNA-N
Synonym 2-methyl-2-phenylsuccinimide, n-desmethylmethsuximide, normesuximide, a-methyl-a-phenylsuccinimide, n-demethylmethsuximide, 2,5-pyrrolidinedione, 3-methyl-3-phenyl, alpha-methyl-alpha-phenylsuccinimide, 3-methyl-3-phenyl-2,5-pyrrolidinedione, .alpha.-methyl-.alpha.-phenylsuccinimide, normethsuximide
PubChem CID 92154
IUPAC Name 3-methyl-3-phenylpyrrolidine-2,5-dione
SMILES CC1(CC(=O)NC1=O)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 189.21
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-phenylsuccinimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.