missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propane-d9-thiol, CDN

Product Code. 30309280
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Product Code. Quantity unitSize
30309280 0.25 g 0.25g
30318039 0.1 g 0.1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30309280

Brand: CDN CDND26440.25G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butanethiol-D9
CAS 99224-24-5
Molecular Formula C4 D9 H S
Quantity 0.25 g
Synonym 2-Propane-1,1,1,3,3,3-d6-thiol, 2-(methyl-d3)- (9 CI), 2-(Methyl-d3)-2-propane-1,1,1,3,3,3-d6-thiol, 1,1-Dimethylethanethiol-d9, tert-Butanethiol-d9, tert-Butyl-d9 mercaptan, tert-Butylthiol-d9, tert-Butanethiol-D9
InChI Formula InChI=1 S/C4H10S/c1-4(2,3)5/h5H,1-3H3/i1D3,2D3,3D3
SMILES [2 H]C([2 H])([2 H])C(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propane-2-thiol
Molecular Weight (g/mol) 99.2427
Formula Weight 99.1068 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.