Learn More
2-Methyl-4-piperazinoquinoline, 97%, Thermo Scientific™
Product Code. 10344152
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10344152 250 mg 250mg
10354152 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10344152 Supplier Thermo Scientific Maybridge Supplier No. RH02136CB
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 82241-22-3
Molecular Formula C14H17N3
Molecular Weight (g/mol) 227.31
MDL Number MFCD01935745
InChI Key XYBLCORUTWKJOI-UHFFFAOYSA-N
Synonym 2-methyl-4-piperazinoquinoline, 2-methyl-4-piperazin-1-yl quinoline, quinoline, 2-methyl-4-1-piperazinyl, 2-methyl-4-quinolyl piperazine, 2-methyl-4-piperazin-1-yl-quinoline, maybridge1_006771, chemdivam_001084, cbmicro_045139, cambridge id 6095001, tos-bb-0571
PubChem CID 702318
SMILES CC1=CC(N2CCNCC2)=C2C=CC=CC2=N1
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 2-methyl-4-piperazin-1-ylquinoline
Formula Weight 227.31
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-piperazinoquinoline