missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzophenone, 98+%

Product Code. 11462497
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11462497

Brand: Thermo Scientific Alfa Aesar A14779.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-58-8 | C14H12O | 196.249 g/mol

2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressure.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-58-8
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008518
InChI Key CKGKXGQVRVAKEA-UHFFFAOYSA-N
Synonym 2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone
PubChem CID 67230
IUPAC Name (2-methylphenyl)-phenylmethanone
SMILES CC1=CC=CC=C1C(=O)C2=CC=CC=C2

Specifications

Melting Point -18°C
Density 1.085
Boiling Point 309°C to 311°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 10 g
Beilstein 2045469
Merck Index 14,7317
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzophenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.