missing translation for 'onlineSavingsMsg'
Learn More

2-Methylmalonamide, 95%, Thermo Scientific Chemicals

Product Code. 11347286
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347286 10 g 10g
11337286 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347286 Supplier Thermo Scientific Alfa Aesar Supplier No. L09203.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-63-9
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00043625
InChI Key CGRKYGKHZOCPSZ-UHFFFAOYSA-N
Synonym 2-methylmalonamide, 2-methylmalonodiamide, methylmalonamide, methyl malonamide, 2-methyl malonamide, acmc-20anpx, propanediamide, 2-methyl
PubChem CID 227804
IUPAC Name 2-methylpropanediamide
SMILES CC(C(N)=O)C(N)=O

Specifications

Melting Point 208°C to 210°C
Quantity 10 g
Beilstein 1756100
Formula Weight 116.12
Percent Purity 95%
Chemical Name or Material 2-Methylmalonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.