missing translation for 'onlineSavingsMsg'
Learn More

2-Methylundecane, TRC

Product Code. 30478849
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478849 50 mg 50mg
30482878 250 mg 250mg
30478277 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478849 Supplier Toronto Research Chemicals Supplier No. TRCM70679050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7045-71-8
Molecular Formula C12H26
Molecular Weight (g/mol) 170.33
IUPAC Name 2-methylundecane
SMILES CCCCCCCCCC(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H26/c1-4-5-6-7-8-9-10-11-12(2)3/h12H,4-11H2,1-3H3
Formula Weight 170.2035
Chemical Name or Material 2-Methylundecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.