missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butylfuran, 98%

Product Code. 11411013
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411013 10 g 10g
11421013 50 g 50g
11431013 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411013 Supplier Thermo Scientific Alfa Aesar Supplier No. A13376.09
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4466-24-4 | C8H12O | 124.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4466-24-4
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00047071
InChI Key NWZIYQNUCXUJJJ-UHFFFAOYSA-N
Synonym furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane
PubChem CID 20534
IUPAC Name 2-butylfuran
SMILES CCCCC1=CC=CO1

Specifications

Density 0.886
Boiling Point 153°C to 154°C
Flash Point 32°C (89°F)
Odor Pleasant
Quantity 10 g
UN Number UN1993
Beilstein 107045
Formula Weight 124.18
Percent Purity 98%
Chemical Name or Material 2-n-Butylfuran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.