missing translation for 'onlineSavingsMsg'
Learn More

2-N,N-Dibenzyl Serine Benzyl Ester-13C3, TRC

Product Code. 30375587
Change view
Click to view available options
Quantity:
1 mg
10 g
Unit Size:
10g
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375587 1 mg 1mg
30375589 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375587 Supplier Toronto Research Chemicals Supplier No. TRCD4186771MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-N,N-Dibenzyl Serine Benzyl Ester-13C3
Molecular Formula 13C3 C21 H25 N O3
Quantity 1 mg
Synonym N,N-Bis(phenylmethyl)serine Phenylmethyl Ester-13C3
InChI Formula InChI=1S/C24H25NO3/c26-18-23(24(27)28-19-22-14-8-3-9-15-22)25(16-20-10-4-1-5-11-20)17-21-12-6-2-7-13-21/h1-15,23,26H,16-19H2/i18+1,23+1,24+1
SMILES O[13CH2][13CH](N(Cc1ccccc1)Cc2ccccc2)[13C](=O)OCc3ccccc3
IUPAC Name benzyl 2-(dibenzylamino)-3-hydroxy(1,2,3-13C3)propanoate
Molecular Weight (g/mol) 378.438
Formula Weight 378.194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.