Learn More
2-N,N-Dibenzyl Serine Benzyl Ester-13C3, TRC
Product Code. 30375589
Change view
Click to view available options
Quantity:
1 mg
10 g
Unit Size:
10g
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375589 10 g 10g
30375587 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30375589 Supplier Toronto Research Chemicals Supplier No. TRCD41867710G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-N,N-Dibenzyl Serine Benzyl Ester-13C3
Molecular Formula 13C3 C21 H25 N O3
Quantity 10 g
Synonym N,N-Bis(phenylmethyl)serine Phenylmethyl Ester-13C3
InChI Formula InChI=1S/C24H25NO3/c26-18-23(24(27)28-19-22-14-8-3-9-15-22)25(16-20-10-4-1-5-11-20)17-21-12-6-2-7-13-21/h1-15,23,26H,16-19H2/i18+1,23+1,24+1
SMILES O[13CH2][13CH](N(Cc1ccccc1)Cc2ccccc2)[13C](=O)OCc3ccccc3
IUPAC Name benzyl 2-(dibenzylamino)-3-hydroxy(1,2,3-13C3)propanoate
Molecular Weight (g/mol) 378.438
Formula Weight 378.194