missing translation for 'onlineSavingsMsg'
Learn More

2-NP-AOZ-d4, TRC

Product Code. 30132806
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132806 25 mg 25mg
17907069 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30132806 Supplier Toronto Research Chemicals Supplier No. TRCN89500225MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1007478-57-0
Molecular Formula C10 2H4 H5 N3 O4
Molecular Weight (g/mol) 239.22
Synonym 2-NP-AOZ D4
IUPAC Name 4,4,5,5-tetradeuterio-3-[(E)-(2-nitrophenyl)methylideneamino]-1,3-oxazolidin-2-one
SMILES [2H]C1([2H])OC(=O)N(\N=C\c2ccccc2[N+](=O)[O-])C1([2H])[2H]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H9N3O4/c14-10-12(5-6-17-10)11-7-8-3-1-2-4-9(8)13(15)16/h1-4,7H,5-6H2/b11-7+/i5D2,6D2
Formula Weight 239.0844
Chemical Name or Material 2-NP-AOZ D4
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.