missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën 2-Propanol CHROMASOLV™

Product Code. 15955510
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
Unit Size:
18 L
45 L
7 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15955510 18 L 18 L
15975510 7 L 7 L
15965510 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15955510 Supplier Riedel-de Haën Supplier No. 3486318L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

2-Propanol CHROMASOLV™ for HPLC and organic synthesis, 99.9%.

  • For HPLC and organic synthesis.
  • High-purity, multipurpose solvents.
  • High lot-to-lot consistency.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Neutral
Melting Point -90°C
Color Colorless
CAS Max % 100.00%
Density 0.784 to 0.786 g/cm3 (20°C)
Quantity 18 L
Flash Point 12°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Linear Formula C3H8O
Vapor Pressure 42 hPa (20°C)
Viscosity 2.5 mPa.s (20°C)
UN Number UN1219
Beilstein 635639
Concentration ≥99.90%
Boiling Point 77°C to 83°C (1.013 hPa)
Percent Purity ≥99.90%
Grade HPLC
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.