missing translation for 'onlineSavingsMsg'
Learn More

2-TEDC, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18074472
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074472 10 mg 10mg
18015862 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074472 Supplier Tocris Bioscience Supplier No. 0645/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5-, 12-, 15-Lipoxygenase inhibitor

Potent inhibitor of 5-, 12-, and 15-lipoxygenase (IC50 values are 0.09, 0.013 and 0.5 μM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132465-10-2
Molecular Formula C16H13NO4S
Molecular Weight (g/mol) 315.343
InChI Key KCGLTUBCAGZLHP-XYOKQWHBSA-N
Synonym 2-tedc, chembl37081, tocris-0645, 2-thiophen-2-yl ethyl 2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, e-2-cyano-3-3,4-dihydroxyphenyl acrylic acid 2-2-thienyl ethyl ester, 2-cyano-3-3,4-dihydroxy-phenyl-acrylic acid 2-thiophen-2-yl-ethyl ester, 2-cyano-3-3,4-dihydroxyphenyl propenoic acid 2-2-thienyl ethyl ester, 2-thiophen-2-ylethyl e-2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, 2-propenoic acid,2-cyano-3-3,4-dihydroxyphenyl-,2-2-thienyl ethyl ester, e-9ci
PubChem CID 6069552
IUPAC Name 2-thiophen-2-ylethyl (E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES C1=CSC(=C1)CCOC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Quantity 10 mg
Formula Weight 315.34
Percent Purity >99%
Chemical Name or Material 2-TEDC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.