Learn More
2'-(Trifluoromethoxy)acetophenone, 98+%, Thermo Scientific™
Product Code. 11399935
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399935 5 g 5g
11389935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11399935 Supplier Thermo Scientific Alfa Aesar Supplier No. L19906.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 220227-93-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00236320
InChI Key XTXSTESGCXKUIH-UHFFFAOYSA-N
Synonym 2'-trifluoromethoxy acetophenone, 1-2-trifluoromethoxy phenyl ethanone, 1-2-trifluoromethoxy phenyl ethan-1-one, 1-acetyl-2-trifluoromethoxy benzene, pubchem4295, 2-trifluoromethoxy acetophenone98, 1-2-trifluoromethoxyphenyl ethanone, 2'-trifluoromethoxy acetophenone98
PubChem CID 2777176
IUPAC Name 1-[2-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=CC=C1OC(F)(F)F
Specifications
Density 1.265
Refractive Index 1.45
Quantity 5 g
Beilstein 2259035
Formula Weight 204.15
Percent Purity ≥98%
Chemical Name or Material 2'-(Trifluoromethoxy)acetophenone