missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 11402287
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402287 5 g 5g
11412287 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402287 Supplier Thermo Scientific Alfa Aesar Supplier No. A14202.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1535-75-7
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00035959
InChI Key ZFCOUBUSGHLCDT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy aniline, benzenamine, 2-trifluoromethoxy, 2-trifluoromethoxy-phenylamine, o-trifluoromethoxy aniline, alpha,alpha,alpha-trifluoro-o-anisidine, o-trifluoromethoxyaniline, 2-trifluoromethoxy aminobenzene, 2-trifluoromethoxy phenylamine, 2-trifluoromethoxy phenyl amine
PubChem CID 73754
IUPAC Name 2-(trifluoromethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC(F)(F)F

Specifications

Density 1.301
Boiling Point 61°C to 63°C (15 mmHg)
Flash Point 63°C (147°F)
Refractive Index 1.462
Quantity 5 g
UN Number UN2810
Beilstein 2803814
Formula Weight 177.13
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.