missing translation for 'onlineSavingsMsg'
Learn More

3-[4-[1-[4-(3-Chloro-2-hydroxypropoxy)phenyl]-1-methylethyl]phenoxy]-1,2-propanediol, TRC

Product Code. 30359166
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359166 10 mg 10mg
30358331 25 mg 25mg
30355985 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359166 Supplier Toronto Research Chemicals Supplier No. TRCC31525010MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 227947-06-0
Molecular Formula C21 H27 Cl O5
Molecular Weight (g/mol) 394.89
Synonym BADGE.H2O.HCl, BADGE.HCl.H2O, 1,2-Propanediol, 3-[4-[1-[4-(3-chloro-2-hydroxypropoxy)phenyl]-1-methylethyl]phenoxy]-, Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether, 3-[4-[1-[4-(3-Chloro-2-hydroxypropoxy)phenyl]-1-methylethyl]phenoxy]-1,2-propanediol
IUPAC Name 3-[4-[2-[4-(3-chloro-2-hydroxypropoxy)phenyl]propan-2-yl]phenoxy]propane-1,2-diol
SMILES CC(C)(c1ccc(OCC(O)CO)cc1)c2ccc(OCC(O)CCl)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H27ClO5/c1-21(2,15-3-7-19(8-4-15)26-13-17(24)11-22)16-5-9-20(10-6-16)27-14-18(25)12-23/h3-10,17-18,23-25H,11-14H2,1-2H3
Formula Weight 394.1547
Chemical Name or Material Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.