missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetonitrile, 97%

Product Code. p-7027469
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411297 5 g 5g
11421297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411297 Supplier Thermo Scientific Alfa Aesar Supplier No. B24247.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13338-63-1 | C11H13NO3 | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13338-63-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00001912
InChI Key ACFJNTXCEQCDBX-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetonitrile, 2-3,4,5-trimethoxyphenyl acetonitrile, 3,4,5-trimethoxybenzyl cyanide, 3,4,5-trimethoxybenzylnitrile, 3,4,5-trimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylacetylnitryl, 3,4,5-trimethoxyphenyl acetonitrile, acetonitrile, 3,4,5-trimethoxyphenyl, 2-3,4,5-trimethoxyphenyl ethanenitrile
PubChem CID 25887
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetonitrile
SMILES COC1=CC(CC#N)=CC(OC)=C1OC

Specifications

Melting Point 75°C to 79°C
Quantity 5 g
UN Number UN3439
Beilstein 2214548
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 3,4,5-Trimethoxyphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.