missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorobenzoyl)acrylic acid, 99%

Product Code. 11337796
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337796 5 g 5g
11347796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337796 Supplier Thermo Scientific Alfa Aesar Supplier No. L11467.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6269-33-6 | C10H7ClO3 | 210.613 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6269-33-6
Molecular Formula C10H7ClO3
Molecular Weight (g/mol) 210.613
MDL Number MFCD00041465
InChI Key VQVQEUFKSRHRCT-AATRIKPKSA-N
Synonym 3-4-chlorobenzoyl acrylic acid, p-chlorobenzoylacrylic acid, trans-3-4-chlorobenzoyl acrylic acid, e-4-4-chlorophenyl-4-oxo-2-butenoic acid, 2e-4-4-chlorophenyl-4-oxobut-2-enoic acid, acide e 4-4-chlorophenyl 4-oxo 2-butenoique french, 2-butenoic acid, 4-4-chlorophenyl-4-oxo-, e, 4-4-chlorophenyl-4-oxo-2-butenoic acid, e-4-4-chlorophenyl-4-oxobut-2-enoic acid, e-4-4-chlorophenyl-4-oxo-but-2-enoic acid
PubChem CID 671716
IUPAC Name (E)-4-(4-chlorophenyl)-4-oxobut-2-enoic acid
SMILES C1=CC(=CC=C1C(=O)C=CC(=O)O)Cl

Specifications

Melting Point 157°C to 159°C
Quantity 5 g
Beilstein 2719881
Formula Weight 210.62
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorobenzoyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.