missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenethyl alcohol, 97%

Product Code. 10159981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10159981 1 g Glass Bottle 1g
10622223 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10159981 Supplier Thermo Scientific Acros Supplier No. 350770010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 35364-79-5 | C8H8Cl2O | 191.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35364-79-5
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.051
MDL Number MFCD00800673
InChI Key GITOMJDYNUMCOV-UHFFFAOYSA-N
Synonym 2-3,4-dichlorophenyl ethanol, 3,4-dichlorophenethyl alcohol, 3,4-dichlorophenethylalcohol, 2-3,4-dichlorophenyl ethan-1-ol, benzeneethanol, 3,4-dichloro, 2-3,4-dichlorophenyl-ethanol, 3,4-dichlorobenzeneethanol, benzeneethanol,3,4-dichloro, acmc-1adlk, 2-3,4-dichloro-phenyl-ethanol
PubChem CID 244558
IUPAC Name 2-(3,4-dichlorophenyl)ethanol
SMILES C1=CC(=C(C=C1CCO)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.5620 to 1.5680 (20°C, 589nm)
Quantity 1 g
Beilstein 06, IV, 3081
Formula Weight 191.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorophenethyl alcohol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorophenethylalcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.