missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%

Product Code. 11452080
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452080 5 g 5g
15457745 250 mg 250mg
11442080 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452080 Supplier Thermo Scientific Alfa Aesar Supplier No. H28557.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1073371-72-8 | C16H25BO3 | 276.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073371-72-8
Molecular Formula C16H25BO3
Molecular Weight (g/mol) 276.183
MDL Number MFCD09953503
InChI Key ZJNJYHQBUIHZLN-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1-propylboronic acid pinacol ester, 2-3-4-methoxyphenyl propyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4-methoxyphenyl propylboronic acid pinacol ester, 3-4-methoxyphenyl propylboronic acid,pinacol ester, 3-4-methoxyphenyl propylboronic acid, pinacol ester
PubChem CID 46739066
IUPAC Name 2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCC2=CC=C(C=C2)OC

Specifications

Quantity 5 g
Formula Weight 276.19
Percent Purity 97%
Chemical Name or Material 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.