missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzylamine, 97%, Thermo Scientific™

Product Code. 11474877
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474877 5 g 5g
11484877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474877 Supplier Thermo Scientific Alfa Aesar Supplier No. B21837.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Piperonylamine is a metabolite of dopamine analogs. It is a pharmaceutical intermediate.

Solubility
Sparingly soluble in water. (0.9 g/100 mL).

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-50-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00005840
InChI Key ZILSBZLQGRBMOR-UHFFFAOYSA-N
Synonym piperonylamine, benzo d 1,3 dioxol-5-ylmethanamine, 3,4-methylenedioxybenzylamine, 1,3-benzodioxol-5-ylmethylamine, 1,3-benzodioxole-5-methanamine, 2h-1,3-benzodioxol-5-ylmethanamine, 3,4-methylenedioxy benzylamine, benzo-1,3-dioxole-5-methylamine, 1-1,3-benzodioxol-5-yl methanamine, 5-aminomethyl-1,3-benzodioxole
PubChem CID 75799
SMILES NCC1=CC=C2OCOC2=C1

Specifications

Density 1.214
Boiling Point 136°C to 137°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.564
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water. (0.9g/100 mL).
IUPAC Name 1,3-benzodioxol-5-ylmethanamine
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 3,4-(Methylenedioxy)benzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.