missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylacetic acid, 97%

Product Code. 11453168
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453168 1 g 1g
11463168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453168 Supplier Thermo Scientific Alfa Aesar Supplier No. A18922.03
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 85068-33-3 | C10H6F6O2 | 272.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-33-3
Molecular Formula C10H6F6O2
Molecular Weight (g/mol) 272.15
MDL Number MFCD00009908
InChI Key PAWSKKHEEYTXSA-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 3,5-bis-trifluoromethylphenylacetic acid, mbt-paa, 2-3,5-di trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl acetic acid, 3,5-di trifluoromethyl phenylacetic acid, 3,5-bis trifluoromethyl phenylaceticacid
PubChem CID 123587
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 6813447
Formula Weight 272.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.