missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-O-p-chlorobenzoyl Floxuridine-13C,15N2, TRC

Product Code. 30378916
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378916 10 mg 10mg
30384053 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378916 Supplier Toronto Research Chemicals Supplier No. TRCD43228210MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Di-O-p-chlorobenzoyl Floxuridine-13C,15N2
Molecular Formula 13C C22 H17 Cl2 F 15N2 O7
Quantity 10 mg
Synonym 2'-Deoxy-5-fluoro-uridine 3',5'-Bis(4-chlorobenzoate)-13C,15N2
InChI Formula InChI=1S/C23H17Cl2FN2O7/c24-14-5-1-12(2-6-14)21(30)33-11-18-17(35-22(31)13-3-7-15(25)8-4-13)9-19(34-18)28-10-16(26)20(29)27-23(28)32/h1-8,10,17-19H,9,11H2,(H,27,29,32)/t17-,18+,19+/m0/s1/i23+1,27+1,28+1
SMILES FC1=C[15N]([C@H]2C[C@H](OC(=O)c3ccc(Cl)cc3)[C@@H](COC(=O)c4ccc(Cl)cc4)O2)[13C](=O)[15NH]C1=O
IUPAC Name [(2R,3S,5R)-3-(4-chlorobenzoyl)oxy-5-(5-fluoro-2,4-dioxo(213C,1,3-15N2)pyrimidin-1-yl)oxolan-2-yl]methyl 4-chlorobenzoate
Molecular Weight (g/mol) 526.274
Formula Weight 525.037
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.