missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-hexyn-3-ol-d5, TRC

Product Code. 30385392
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385392 2.5 mg 2.5mg
30379320 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30385392 Supplier Toronto Research Chemicals Supplier No. TRCD4765922.5MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dimethyl-1-hexyn-3-ol-d5
Molecular Formula C8 D5 H9 O
Quantity 2.5 mg
Synonym 3,5-Dimethylhexyn-3-ol-d5, NSC 978-d5, Surfynol 61-d5
InChI Formula InChI=1S/C8H14O/c1-5-8(4,9)6-7(2)3/h1,7,9H,6H2,2-4H3/i4D3,6D2
SMILES [2H]C([2H])([2H])C(O)(C#C)C([2H])([2H])C(C)C
IUPAC Name 4,4-dideuterio-5-methyl-3-(trideuteriomethyl)hex-1-yn-3-ol
Molecular Weight (g/mol) 131.227
Formula Weight 131.136
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.