missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-adamantanol, 98+%

Product Code. 15421209
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421209 100 g 100g
15401209 5 g 5g
15411209 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421209 Supplier Thermo Scientific Alfa Aesar Supplier No. H64137.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 702-82-9 | C10H17NO | 167.252 g/mol

It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

Solubility
Soluble in organic solvents,insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 702-82-9
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD01821204
InChI Key DWPIPTNBOVJYAD-UHFFFAOYSA-N
Synonym 3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
PubChem CID 658645
IUPAC Name 3-aminoadamantan-1-ol
SMILES C1C2CC3(CC1CC(C2)(C3)O)N

Specifications

Melting Point 265°C (literature)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2802127
Solubility Information Soluble in organic solvents,insoluble in water.
Formula Weight 167.25
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 3-Amino-1-adamantanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.