missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific™

Product Code. 10050640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10050640 250 g Glass bottle 250g
10783003 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10050640 Supplier Thermo Scientific Acros Supplier No. 103982500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2458-12-0
MDL Number MFCD00007742
InChI Key XKFIFYROMAAUDL-UHFFFAOYSA-N
Synonym 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh
PubChem CID 75568
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 164.0°C to 168.0°C
Color Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9NO2
Linear Formula H2NC6H3(CH3)CO2H
Quantity 250 g
Beilstein 14, 487
Molecular Weight (g/mol) 151.165
Formula Weight 151.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylbenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.