missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 10413785 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
25 g
Unit Size:
25g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10413785 25 g 25g
10783973 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10413785 Supplier Thermo Scientific Acros Supplier No. 358860250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156682-54-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD02093063
InChI Key WIJNYNBSPQMJGO-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
PubChem CID 2734313
IUPAC Name [3-(benzyloxy)phenyl]boronic acid
SMILES OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 25 g
Formula Weight 228.05
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-(Benzyloxy)phenylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.