missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-propanol, TRC

Product Code. 30133599
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133599 10 g 10g
30011968 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133599 Supplier Toronto Research Chemicals Supplier No. TRCB68665010G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3 H7 Br O
Molecular Weight (g/mol) 138.99
Synonym 1-Propanol, 3-bromo- (6CI, 7CI, 8CI, 9CI, ACI), 3-Bromo-1-propanol (ACI), 1-Bromo-3-hydroxypropane, 1-Bromo-3-propanol, 3-Bromopropane-1-ol, 3-Bromopropyl alcohol, 3-Hydroxypropyl bromide, Trimethylene bromohydrin
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H7BrO/c4-2-1-3-5/h5H,1-3H2
Formula Weight 137.968
Chemical Name or Material 3-Bromo-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.